Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARID3A
Entrez Gene ID
511283
Gene Name
AT-rich interaction domain 3A
Gene Aliases
-
Chromosome Location
Chr.7: 43465805 - 43498751 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205695.1 NP_001192624.1 6 - 7 1490 86

View Full Product Details