Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNX33
Entrez Gene ID
511561
Gene Name
sorting nexin 33
Gene Aliases
SH3PX3
Chromosome Location
Chr.21: 33233415 - 33241926 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205702.1 NP_001192631.1 1 - 2 1460 63

View Full Product Details