Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COLEC10
Entrez Gene ID
512030
Gene Name
collectin subfamily member 10
Gene Aliases
-
Chromosome Location
Chr.14: 45105393 - 45281289 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205713.1 NP_001192642.1 5 - 6 445 96

View Full Product Details