Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB24
Entrez Gene ID
539966
Gene Name
zinc finger and BTB domain containing 24
Gene Aliases
-
Chromosome Location
Chr.9: 40680757 - 40691831 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205737.1 NP_001192666.1 1 - 2 955 120

View Full Product Details