Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPTY2D1
Entrez Gene ID
539967
Gene Name
SPT2 chromatin protein domain containing 1
Gene Aliases
-
Chromosome Location
Chr.29: 26103453 - 26119994 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205738.1 NP_001192667.1 1 - 2 79 52

View Full Product Details