Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATN1
Entrez Gene ID
513125
Gene Name
atrophin 1
Gene Aliases
-
Chromosome Location
Chr.5: 103521298 - 103533994 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205748.2 NP_001192677.1 5 - 6 2630 71
GenBank mRNA BC149148.1 - 1 - 2 798 71

View Full Product Details