Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM15
Entrez Gene ID
540074
Gene Name
tripartite motif containing 15
Gene Aliases
-
Chromosome Location
Chr.23: 28802708 - 28815062 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205764.1 NP_001192693.1 2 - 3 456 63

View Full Product Details