Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM21A
Entrez Gene ID
513740
Gene Name
family with sequence similarity 21, member A
Gene Aliases
-
Chromosome Location
Chr.28: 44112887 - 44157819 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205767.2 NP_001192696.1 13 - 14 1275 82

View Full Product Details