Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM21A
Entrez Gene ID
513740
Gene Name
family with sequence similarity 21, member A
Gene Aliases
-
Chromosome Location
Chr.28: 44112887 - 44157819 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205767.2 NP_001192696.1 30 - 31 3962 91

View Full Product Details