Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A47
Entrez Gene ID
516483
Gene Name
solute carrier family 25 member 47
Gene Aliases
-
Chromosome Location
Chr.21: 65228286 - 65233524 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205829.2 NP_001192758.2 1 - 2 31 87

View Full Product Details