Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AQP9
Entrez Gene ID
516762
Gene Name
aquaporin 9
Gene Aliases
-
Chromosome Location
Chr.10: 52087959 - 52136244 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205833.3 NP_001192762.2 3 - 4 371 65

View Full Product Details