Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SGMS2
Entrez Gene ID
520954
Gene Name
sphingomyelin synthase 2
Gene Aliases
-
Chromosome Location
Chr.6: 17292878 - 17392257 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205877.3 NP_001192806.1 1 - 2 453 114

View Full Product Details