Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC30A1
Entrez Gene ID
522265
Gene Name
solute carrier family 30 member 1
Gene Aliases
-
Chromosome Location
Chr.16: 71867915 - 71872152 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205893.2 NP_001192822.1 1 - 2 617 79

View Full Product Details