Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNNM4
Entrez Gene ID
522382
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 4
Gene Aliases
-
Chromosome Location
Chr.11: 2722661 - 2760313 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205896.2 NP_001192825.2 5 - 6 1958 122

View Full Product Details