Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CNNM4
Entrez Gene ID
522382
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 4
Gene Aliases
-
Chromosome Location
Chr.11: 2722661 - 2760313 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205896.2 NP_001192825.2 6 - 7 2142 63

View Full Product Details