Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TESMIN
Entrez Gene ID
523398
Gene Name
testis expressed metallothionein like protein
Gene Aliases
-
Chromosome Location
Chr.29: 46118488 - 46159248 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205903.3 NP_001192832.1 2 - 3 577 75

View Full Product Details