Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PMS2
Entrez Gene ID
527039
Gene Name
PMS1 homolog 2, mismatch repair system component
Gene Aliases
-
Chromosome Location
Chr.25: 38006833 - 38030341 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205938.1 NP_001192867.1 10 - 11 1141 107

View Full Product Details