Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD50
Entrez Gene ID
527956
Gene Name
ankyrin repeat domain 50
Gene Aliases
-
Chromosome Location
Chr.17: 33129811 - 33170654 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205949.2 NP_001192878.1 2 - 3 741 76

View Full Product Details