Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GZMK
Entrez Gene ID
528871
Gene Name
granzyme K
Gene Aliases
-
Chromosome Location
Chr.20: 24073111 - 24083866 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205959.1 NP_001192888.1 1 - 2 116 75

View Full Product Details