Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYT15
Entrez Gene ID
529391
Gene Name
synaptotagmin 15
Gene Aliases
-
Chromosome Location
Chr.28: 41777230 - 41786760 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205963.2 NP_001192892.2 6 - 7 979 60

View Full Product Details