Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INHBC
Entrez Gene ID
530785
Gene Name
inhibin subunit beta C
Gene Aliases
-
Chromosome Location
Chr.5: 56011447 - 56021810 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205983.1 NP_001192912.1 1 - 2 315 79

View Full Product Details