Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLK9
Entrez Gene ID
530795
Gene Name
kallikrein related peptidase 9
Gene Aliases
KLNF
Chromosome Location
Chr.18: 56980171 - 56986261 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205984.3 NP_001192913.3 4 - 5 605 100

View Full Product Details