Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCL3
Entrez Gene ID
531942
Gene Name
BCL3 transcription coactivator
Gene Aliases
-
Chromosome Location
Chr.18: 52520139 - 52529472 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205993.2 NP_001192922.1 2 - 3 502 72

View Full Product Details