Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BMP7
Entrez Gene ID
540595
Gene Name
bone morphogenetic protein 7
Gene Aliases
-
Chromosome Location
Chr.13: 58889622 - 58975046 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206015.1 NP_001192944.1 3 - 4 1235 68

View Full Product Details