Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OTX1
Entrez Gene ID
540628
Gene Name
orthodenticle homeobox 1
Gene Aliases
-
Chromosome Location
Chr.11: 61541979 - 61551633 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206017.1 NP_001192946.1 1 - 2 103 89

View Full Product Details