Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CATSPER3
Entrez Gene ID
540820
Gene Name
cation channel sperm associated 3
Gene Aliases
-
Chromosome Location
Chr.7: 46421402 - 46457384 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206032.1 NP_001192961.1 4 - 5 678 98

View Full Product Details