Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LEFTY2
Entrez Gene ID
534018
Gene Name
left-right determination factor 2
Gene Aliases
-
Chromosome Location
Chr.16: 28910505 - 28913354 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206043.2 NP_001192972.2 2 - 3 496 48

View Full Product Details