Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DDX50
Entrez Gene ID
534331
Gene Name
DExD-box helicase 50
Gene Aliases
-
Chromosome Location
Chr.28: 25178284 - 25211518 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206053.2 NP_001192982.1 2 - 3 511 107

View Full Product Details