Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
E2F1
Entrez Gene ID
535369
Gene Name
E2F transcription factor 1
Gene Aliases
-
Chromosome Location
Chr.13: 63129139 - 63138330 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206079.1 NP_001193008.1 2 - 3 358 110

View Full Product Details