Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CALN1
Entrez Gene ID
538163
Gene Name
calneuron 1
Gene Aliases
-
Chromosome Location
Chr.25: 28636197 - 29098356 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206115.1 NP_001193044.1 5 - 6 664 76

View Full Product Details