Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GNMT
Entrez Gene ID
538212
Gene Name
glycine N-methyltransferase
Gene Aliases
-
Chromosome Location
Chr.23: 16571898 - 16575330 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206116.1 NP_001193045.1 4 - 5 611 80

View Full Product Details