Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C3H1orf87
Entrez Gene ID
504762
Gene Name
chromosome 3 C1orf87 homolog
Gene Aliases
-
Chromosome Location
Chr.3: 85704935 - 85786705 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206121.3 NP_001193050.3 5 - 6 810 106
GenBank mRNA BC149431.1 - 5 - 6 817 106

View Full Product Details