Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTP4A1
Entrez Gene ID
613326
Gene Name
protein tyrosine phosphatase 4A1
Gene Aliases
-
Chromosome Location
Chr.9: 448438 - 458078 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206124.2 NP_001193053.1 3 - 4 857 90
NM_001354592.1 NP_001341521.1 3 - 4 831 90
GenBank mRNA BC109916.1 - 3 - 4 719 90
EH374978.1 - 2 - 3 446 90

View Full Product Details