Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MIEF1
Entrez Gene ID
613306
Gene Name
mitochondrial elongation factor 1
Gene Aliases
SMCR7L
Chromosome Location
Chr.5: 110928089 - 110943940 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206126.1 NP_001193055.1 2 - 3 319 102

View Full Product Details