Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB5B
Entrez Gene ID
539150
Gene Name
RAB5B, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.5: 57321742 - 57337445 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206191.2 NP_001193120.1 1 - 2 34 79

View Full Product Details