Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC5A
Entrez Gene ID
615201
Gene Name
C-type lectin domain containing 5A
Gene Aliases
-
Chromosome Location
Chr.4: 105219594 - 105245332 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206203.1 NP_001193132.1 1 - 2 78 95

View Full Product Details