Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RSBN1
Entrez Gene ID
508231
Gene Name
round spermatid basic protein 1
Gene Aliases
-
Chromosome Location
Chr.3: 29579106 - 29618905 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206227.1 NP_001193156.1 3 - 4 1585 91

View Full Product Details