Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCDH20
Entrez Gene ID
616023
Gene Name
protocadherin 20
Gene Aliases
-
Chromosome Location
Chr.12: 477004 - 480875 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206231.1 NP_001193160.1 1 - 2 135 76

View Full Product Details