Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C19H17orf98
Entrez Gene ID
616773
Gene Name
chromosome 19 C17orf98 homolog
Gene Aliases
-
Chromosome Location
Chr.19: 39432066 - 39435348 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206258.1 NP_001193187.1 2 - 3 305 64

View Full Product Details