Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FA2H
Entrez Gene ID
516891
Gene Name
fatty acid 2-hydroxylase
Gene Aliases
-
Chromosome Location
Chr.18: 2151163 - 2205766 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192455.1 NP_001179384.1 6 - 7 1087 85

View Full Product Details