Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZSWIM2
Entrez Gene ID
517320
Gene Name
zinc finger SWIM-type containing 2
Gene Aliases
-
Chromosome Location
Chr.2: 9491627 - 9512833 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192467.1 NP_001179396.1 8 - 9 1104 81

View Full Product Details