Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LCTL
Entrez Gene ID
518599
Gene Name
lactase like
Gene Aliases
-
Chromosome Location
Chr.10: 13404231 - 13420411 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192493.3 NP_001179422.1 12 - 13 1591 89

View Full Product Details