Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YTHDF3
Entrez Gene ID
519708
Gene Name
YTH N6-methyladenosine RNA binding protein 3
Gene Aliases
-
Chromosome Location
Chr.14: 28141086 - 28174571 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192513.3 NP_001179442.1 3 - 4 381 117

View Full Product Details