Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLSCR5
Entrez Gene ID
521251
Gene Name
phospholipid scramblase family member 5
Gene Aliases
-
Chromosome Location
Chr.1: 121852641 - 121878383 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192535.2 NP_001179464.2 1 - 2 174 89

View Full Product Details