Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNRPB2
Entrez Gene ID
767873
Gene Name
small nuclear ribonucleoprotein polypeptide B2
Gene Aliases
-
Chromosome Location
Chr.13: 10562583 - 10576213 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192552.1 NP_001179481.1 2 - 3 147 112
GenBank mRNA BC102328.1 - 2 - 3 146 112

View Full Product Details