Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCTEX1D1
Entrez Gene ID
522474
Gene Name
Tctex1 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.3: 78541136 - 78570775 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192562.1 NP_001179491.1 3 - 4 252 105

View Full Product Details