Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FCRLB
Entrez Gene ID
522702
Gene Name
Fc receptor like B
Gene Aliases
-
Chromosome Location
Chr.3: 7884632 - 7889585 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192565.1 NP_001179494.1 3 - 4 310 59

View Full Product Details