Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC3H12B
Entrez Gene ID
524199
Gene Name
zinc finger CCCH-type containing 12B
Gene Aliases
-
Chromosome Location
Chr.X: 94978170 - 95054375 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192594.2 NP_001179523.2 2 - 3 744 102

View Full Product Details