Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM20
Entrez Gene ID
525419
Gene Name
RNA binding motif protein 20
Gene Aliases
-
Chromosome Location
Chr.26: 31199404 - 31399463 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192613.3 NP_001179542.3 9 - 10 2499 71

View Full Product Details