Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPAT2
Entrez Gene ID
526739
Gene Name
glycerol-3-phosphate acyltransferase 2, mitochondrial
Gene Aliases
-
Chromosome Location
Chr.11: 2164620 - 2176621 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192634.2 NP_001179563.2 17 - 18 2023 91

View Full Product Details